This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04655038_m1
See other DGAT2 GE Assays ›
Gene Symbol:
DGAT2
Gene Name:
diacylglycerol O-acyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 27720878 - 27753735 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04655038_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DGAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001314305.1 NP_001301234.1 5 - 6 564 57
XM_018058853.1 XP_017914342.1 5 - 6 561 57
GenBank mRNA HM566448.1 - 5 - 6 565 57
KA327728.1 - 5 - 6 748 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0