This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04656592_m1
See other BCAT1 GE Assays ›
Gene Symbol:
BCAT1
Gene Name:
branched chain amino acid transaminase 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 83672357 - 83789003 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04656592_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013964159.2 XP_013819613.1 3 - 4 560 61
XM_018048393.1 XP_017903882.1 3 - 4 603 61
XM_018048394.1 XP_017903883.1 3 - 4 457 61
XM_018048395.1 XP_017903884.1 3 - 4 914 61
GenBank mRNA JO640661.2 - 8 - 9 1195 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline