This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04660102_m1
See other ACMSD GE Assays ›
Gene Symbol:
ACMSD
Gene Name:
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.2: 73805813 - 73869002 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04660102_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACMSD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005676200.3 XP_005676257.2 2 - 3 260 61
XM_013969990.2 XP_013825444.2 3 - 4 558 61
XM_013969992.2 XP_013825446.2 3 - 4 559 61
XM_013969997.2 XP_013825451.1 3 - 4 559 61
XM_013969999.2 XP_013825453.1 3 - 4 559 61
XM_013970003.2 XP_013825457.2 3 - 4 534 61
GenBank mRNA JO434414.2 - 2 - 3 267 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

decarboxylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0