This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04661213_m1
See other GCNT7 GE Assays ›
Gene Symbol:
GCNT7
Gene Name:
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases:
-
Chromosome Location:
Chr.13: 58749847 - 58761270 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04661213_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005688318.3 XP_005688375.1 2 - 3 2611 115
XM_013968770.2 XP_013824224.1 1 - 2 1965 115
GenBank mRNA JO574451.2 - 2 - 3 235 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0