This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04663287_g1
See other CLPB GE Assays ›
Gene Symbol:
CLPB
Gene Name:
ClpB homolog, mitochondrial AAA ATPase chaperonin
Gene Aliases:
-
Chromosome Location:
Chr.15: 31410695 - 31553754 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04663287_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013969731.2 XP_013825185.2 3 - 4 751 55
XM_013969732.2 XP_013825186.2 3 - 4 600 55
XM_018059407.1 XP_017914896.1 1 - 2 99 55
GenBank mRNA KA348712.1 - 3 - 4 580 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0