Assay Details
| Gene Symbol: CDH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057599.1 | XP_017913088.1 | 10 - 11 | 1737 | 54 | |
| GenBank mRNA | KA347273.1 | - | 10 - 11 | 1625 | 54 | |
More Information
Set Membership: |
|

CDH4
Genomic Map