This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04667004_m1
See other CADM1 GE Assays ›
Gene Symbol:
CADM1
Gene Name:
cell adhesion molecule 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 56277080 - 56634777 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04667004_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CADM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013969498.2 XP_013824952.2 1 - 2 253 67
XM_018059752.1 XP_017915241.1 1 - 2 253 67
XM_018059753.1 XP_017915242.1 1 - 2 253 67
XM_018059754.1 XP_017915243.1 1 - 2 253 67
XM_018059755.1 XP_017915244.1 1 - 2 159 67
XM_018059756.1 XP_017915245.1 1 - 2 253 67
GenBank mRNA JO560530.2 - 10 - 11 1435 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0