This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04669447_m1
See other PITPNC1 GE Assays ›
Gene Symbol:
PITPNC1
Gene Name:
phosphatidylinositol transfer protein cytoplasmic 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 48794971 - 49056513 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04669447_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITPNC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005694019.3 XP_005694076.2 2 - 3 1018 64
XM_013972268.2 XP_013827722.1 2 - 3 1021 64
XM_018065380.1 XP_017920869.1 2 - 3 1001 64
XM_018065381.1 XP_017920870.1 3 - 4 351 64
XM_018065382.1 XP_017920871.1 2 - 3 238 64
XM_018065383.1 XP_017920872.1 2 - 3 1007 64
GenBank mRNA JO623471.2 - 3 - 4 913 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0