This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04676096_m1
See other PRSS2 GE Assays ›
Gene Symbol:
PRSS2
Gene Name:
serine protease 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 14549605 - 14553632 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04676096_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005679544.3 XP_005679601.3 3 - 4 671 83
GenBank mRNA JT688573.1 - 2 - 3 389 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0