This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04677467_m1
See other IGSF6 GE Assays ›
Gene Symbol:
IGSF6
Gene Name:
immunoglobulin superfamily member 6
Gene Aliases:
-
Chromosome Location:
Chr.25: 20076816 - 20087660 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04677467_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005697599.3 XP_005697656.1 1 - 2 554 71
GenBank mRNA JO330658.2 - 5 - 6 1761 71
JO594298.2 - 5 - 6 1774 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t6srt:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0