This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04677480_m1
See other IL4R GE Assays ›
Gene Symbol:
IL4R
Gene Name:
interleukin 4 receptor
Gene Aliases:
-
Chromosome Location:
Chr.25: 24784671 - 24834755 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04677480_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL4R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005697626.3 XP_005697683.2 9 - 10 1021 61
XM_018040645.1 XP_017896134.1 9 - 10 1041 61
XM_018040646.1 XP_017896135.1 9 - 10 1044 61
XM_018040647.1 XP_017896136.1 10 - 11 1211 61
XM_018040648.1 XP_017896137.1 10 - 11 1214 61
XM_018040649.1 XP_017896138.1 7 - 8 788 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0