This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04677497_m1
See other ORAI2 GE Assays ›
Gene Symbol:
ORAI2
Gene Name:
ORAI calcium release-activated calcium modulator 2
Gene Aliases:
-
Chromosome Location:
Chr.25: 35508693 - 35523123 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04677497_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORAI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005697805.3 XP_005697862.1 2 - 3 1395 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

ion channel transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0