This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04677785_m1
See other VSIG4 GE Assays ›
Gene Symbol:
VSIG4
Gene Name:
V-set and immunoglobulin domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.X|NW_017189516: 21109230 - 21154374 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04677785_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005700851.3 XP_005700908.1 1 - 2 132 68
GenBank mRNA JT762180.1 - 1 - 2 159 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0