This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04679931_m1
See other GTF2E1 GE Assays ›
Gene Symbol:
GTF2E1
Gene Name:
general transcription factor IIE subunit 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 65117181 - 65151714 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04679931_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005674994.3 XP_005675051.1 4 - 5 1061 89
XM_018043725.1 XP_017899214.1 4 - 5 1088 89
XM_018043773.1 XP_017899262.1 4 - 5 1466 89
XM_018043823.1 XP_017899312.1 4 - 5 1481 89
GenBank mRNA JO321830.2 - 1 - 2 468 89
JO416305.2 - 1 - 2 437 89
JO416306.2 - 1 - 2 437 89
JO445464.2 - 1 - 2 465 89
JO594884.2 - 1 - 2 1750 89
JO619630.2 - 1 - 2 481 89
KA321199.1 - 4 - 5 987 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0