This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04680741_g1
See other TMEM173 GE Assays ›
Gene Symbol:
TMEM173
Gene Name:
transmembrane protein 173
Gene Aliases:
STING
Chromosome Location:
Chr.7: 59759509 - 59765655 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04680741_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM173
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319278.1 NP_001306207.1 5 - 6 815 69
XM_005683020.3 XP_005683077.1 6 - 7 929 69
XM_005683021.3 XP_005683078.1 6 - 7 1054 69
XM_005683022.3 XP_005683079.1 6 - 7 925 69
XM_005683023.3 XP_005683080.1 6 - 7 990 69
XM_018049970.1 XP_017905459.1 4 - 5 1265 69
GenBank mRNA JO341443.2 - 3 - 4 1449 69
JO341445.2 - 3 - 4 1449 69
KR060015.1 - 5 - 6 815 69
KT013268.1 - 5 - 6 815 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline