This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04680793_m1
See other SLCO6A1 GE Assays ›
Gene Symbol:
SLCO6A1
Gene Name:
solute carrier organic anion transporter family member 6A1
Gene Aliases:
-
Chromosome Location:
Chr.7: 9124263 - 9219243 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04680793_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO6A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018051637.1 XP_017907126.1 7 - 8 1127 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline