This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04684100_m1
See other AKR1A1 GE Assays ›
Gene Symbol:
AKR1A1
Gene Name:
aldo-keto reductase family 1 member A1
Gene Aliases:
-
Chromosome Location:
Chr.3: 20260827 - 20271462 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04684100_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018043616.1 XP_017899105.1 7 - 8 988 61
XM_018043619.1 XP_017899108.1 8 - 9 1139 61
XM_018043621.1 XP_017899110.1 7 - 8 939 61
GenBank mRNA JO419570.2 - 2 - 3 229 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0