Assay Details
| Gene Symbol: CD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063294.1 | XP_017918783.1 | 7 - 8 | 1653 | 61 | |
| XM_018063295.1 | XP_017918784.1 | 7 - 8 | 1554 | 61 | ||
| XM_018063296.1 | XP_017918785.1 | 5 - 6 | 1128 | 61 | ||
More Information
Set Membership: |
|

CD22
Genomic Map