This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04688236_m1
See other ATP5F1D GE Assays ›
Gene Symbol:
ATP5F1D
Gene Name:
ATP synthase F1 subunit delta
Gene Aliases:
ATP5D
Chromosome Location:
Chr.7: 66797813 - 66799987 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04688236_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5F1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005682906.3 XP_005682963.1 2 - 3 426 65
GenBank mRNA JT763352.1 - 2 - 3 602 65
KA340689.1 - 2 - 3 428 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline