This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04688818_m1
See other DFFB GE Assays ›
Gene Symbol:
DFFB
Gene Name:
DNA fragmentation factor subunit beta
Gene Aliases:
-
Chromosome Location:
Chr.16: 47780509 - 47794298 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04688818_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DFFB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005690758.3 XP_005690815.2 4 - 5 1190 90
XM_005690759.3 XP_005690816.2 6 - 7 957 90
XM_013970263.2 XP_013825717.2 7 - 8 1030 90
GenBank mRNA JO595899.2 - 1 - 2 1143 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0