This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04693378_m1
See other AWAT1 GE Assays ›
Gene Symbol:
AWAT1
Gene Name:
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.X|NW_017189516: 11348657 - 11355476 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04693378_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AWAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005700705.3 XP_005700762.1 2 - 3 284 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0