This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04696582_m1
See other NTMT1 GE Assays ›
Gene Symbol:
NTMT1
Gene Name:
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 99496513 - 99504269 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04696582_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005687265.3 XP_005687322.1 3 - 4 632 71
XM_013968035.2 XP_013823489.1 2 - 3 339 71
XM_018055952.1 XP_017911441.1 3 - 4 625 71
XM_018055953.1 XP_017911442.1 3 - 4 489 71
GenBank mRNA JO638235.2 - 1 - 2 573 71
KA350313.1 - 4 - 5 629 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline