This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04697387_m1
See other HOGA1 GE Assays ›
Gene Symbol:
HOGA1
Gene Name:
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases:
-
Chromosome Location:
Chr.26: 32824911 - 32847626 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04697387_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HOGA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005698298.3 XP_005698355.1 6 - 7 1223 92
XM_018041492.1 XP_017896981.1 2 - 3 738 92
GenBank mRNA JO428160.1 - 1 - 2 1345 92
JO645229.2 - 6 - 7 1226 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0