Assay Details
| Gene Symbol: CHST15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041479.1 | XP_017896968.1 | 6 - 7 | 2050 | 60 | |
| XM_018041480.1 | XP_017896969.1 | 8 - 9 | 2275 | 60 | ||
| XM_018041481.1 | XP_017896970.1 | 7 - 8 | 2133 | 60 | ||
| XM_018041482.1 | XP_017896971.1 | 6 - 7 | 1927 | 60 | ||
| XM_018041483.1 | XP_017896972.1 | 7 - 8 | 2192 | 60 | ||
| XM_018041484.1 | XP_017896973.1 | 6 - 7 | 2326 | 60 | ||
More Information
Set Membership: |
|

CHST15
Genomic Map