This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04698729_g1
See other MAMSTR GE Assays ›
Gene Symbol:
MAMSTR
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.18: 56373046 - 56378785 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04698729_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAMSTR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018062734.1 XP_017918223.1 2 - 3 1445 68
XM_018062735.1 XP_017918224.1 2 - 3 1415 68
XM_018062736.1 XP_017918225.1 4 - 5 378 68
XM_018062737.1 XP_017918226.1 4 - 5 348 68
GenBank mRNA JO546367.2 - 3 - 4 252 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0