This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04700238_g1
See other BAAT GE Assays ›
Gene Symbol:
BAAT
Gene Name:
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.8: 90845021 - 90861304 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04700238_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005684245.3 XP_005684302.1 3 - 4 797 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0