This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04702219_m1
See other EEF1AKMT1 GE Assays ›
Gene Symbol:
EEF1AKMT1
Gene Name:
EEF1A lysine methyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 50933692 - 50957111 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04702219_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1AKMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013968196.2 XP_013823650.2 3 - 4 1000 69
XM_018056541.1 XP_017912030.1 3 - 4 1000 69
XM_018056542.1 XP_017912031.1 4 - 5 728 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline