This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04704700_m1
See other GMNN GE Assays ›
Gene Symbol:
GMNN
Gene Name:
geminin DNA replication inhibitor
Gene Aliases:
-
Chromosome Location:
Chr.23: 16951732 - 16962395 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04704700_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GMNN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005696777.3 XP_005696834.1 4 - 5 507 74
XM_005696778.3 XP_005696835.1 4 - 5 468 74
XM_005696779.3 XP_005696836.1 4 - 5 486 74
XM_013974108.2 XP_013829562.1 4 - 5 525 74
XM_013974110.2 XP_013829564.1 4 - 5 530 74
GenBank mRNA JI861636.1 - 3 - 4 278 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0