This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04707995_g1
See other IL37 GE Assays ›
Gene Symbol:
IL37
Gene Name:
interleukin 37
Gene Aliases:
-
Chromosome Location:
Chr.11: 46246080 - 46257477 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04707995_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686667.3 XP_005686724.2 3 - 4 289 97
XM_005686668.3 XP_005686725.2 3 - 4 289 97
XM_005686669.2 XP_005686726.2 3 - 4 279 97
XM_013967721.2 XP_013823175.2 3 - 4 279 97
GenBank mRNA KA329633.1 - 3 - 4 792 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

interleukin superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0