This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04708108_g1
See other NUDT5 GE Assays ›
Gene Symbol:
NUDT5
Gene Name:
nudix hydrolase 5
Gene Aliases:
-
Chromosome Location:
Chr.13: 16137247 - 16158476 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04708108_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005687856.3 XP_005687913.1 6 - 7 535 58
XM_013968437.2 XP_013823891.1 5 - 6 878 58
XM_018057009.1 XP_017912498.1 6 - 7 610 58
XM_018057010.1 XP_017912499.1 6 - 7 615 58
XM_018057011.1 XP_017912500.1 6 - 7 604 58
GenBank mRNA JT681663.1 - 6 - 7 576 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

pyrophosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0