This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04708528_m1
See other ADGRG5 GE Assays ›
Gene Symbol:
ADGRG5
Gene Name:
adhesion G protein-coupled receptor G5
Gene Aliases:
GPR114
Chromosome Location:
Chr.18: 26875360 - 26902253 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04708528_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005692027.3 XP_005692084.1 4 - 5 503 60
XM_013971049.2 XP_013826503.1 5 - 6 555 60
XM_018062044.1 XP_017917533.1 4 - 5 666 60
GenBank mRNA JO595572.2 - 5 - 6 563 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0