This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04711235_m1
See other HACD2 GE Assays ›
Gene Symbol:
HACD2
Gene Name:
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases:
PTPLB
Chromosome Location:
Chr.1: 67627468 - 67719192 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04711235_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018045763.1 XP_017901252.1 5 - 6 578 75
GenBank mRNA JO336947.2 - 5 - 6 423 75
JO348898.2 - 2 - 3 320 75
JO348899.2 - 2 - 3 320 75
JO409280.2 - 5 - 6 549 75
JO409281.2 - 5 - 6 549 75
JO442515.2 - 5 - 6 574 75
JO453655.2 - 5 - 6 574 75
JO510788.2 - 5 - 6 541 75
JO558922.1 - 2 - 3 367 75
JO574671.2 - 4 - 5 262 75
JO605359.2 - 2 - 3 313 75
JO639335.2 - 1 - 2 244 75
JO639336.2 - 1 - 2 244 75
JT705841.1 - 3 - 4 507 75
KA350340.1 - 5 - 6 568 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0