This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04711423_m1
See other ECM1 GE Assays ›
Gene Symbol:
ECM1
Gene Name:
extracellular matrix protein 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 99976658 - 99982393 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04711423_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005677667.3 XP_005677724.2 1 - 2 563 72
XM_005677668.3 XP_005677725.2 1 - 2 563 72
XM_013962438.2 XP_013817892.2 1 - 2 563 72
XM_013962439.2 XP_013817893.2 1 - 2 563 72
XM_013962440.2 XP_013817894.2 1 - 2 563 72
XM_013962441.2 XP_013817895.2 1 - 2 563 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0