This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04714876_m1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
-
Chromosome Location:
Chr.23: 9524172 - 9945957 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04714876_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018038742.1 XP_017894231.1 9 - 10 4438 71
XM_018038743.1 XP_017894232.1 9 - 10 3473 71
XM_018038744.1 XP_017894233.1 9 - 10 3488 71
XM_018038745.1 XP_017894234.1 8 - 9 3408 71
XM_018038746.1 XP_017894235.1 8 - 9 4196 71
XM_018038747.1 XP_017894236.1 8 - 9 3938 71
XM_018038748.1 XP_017894237.1 8 - 9 3632 71
XM_018038749.1 XP_017894238.1 7 - 8 3489 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline