This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04718314_m1
See other ABCG8 GE Assays ›
Gene Symbol:
ABCG8
Gene Name:
ATP binding cassette subfamily G member 8
Gene Aliases:
-
Chromosome Location:
Chr.11: 25943455 - 25962050 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04718314_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013967599.2 XP_013823053.2 8 - 9 1286 60
XM_013967600.2 XP_013823054.2 8 - 9 1283 60
GenBank mRNA JO419790.2 - 5 - 6 2031 60
JO419791.2 - 5 - 6 2031 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0