Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697552.3 | XP_005697609.2 | 13 - 14 | 1787 | 73 | |
| XM_005697553.3 | XP_005697610.2 | 13 - 14 | 1749 | 73 | ||
| XM_005697554.3 | XP_005697611.2 | 12 - 13 | 1635 | 73 | ||
| XM_018041024.1 | XP_017896513.1 | 12 - 13 | 1635 | 73 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map