Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692633.3 | XP_005692690.1 | 4 - 5 | 1502 | 69 | |
| GenBank mRNA | JT702972.1 | - | 4 - 5 | 1863 | 69 | |
| KA345379.1 | - | 1 - 2 | 359 | 69 | ||
More Information
Set Membership: |
|

ZNF180
Genomic Map