Assay Details
| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681787.3 | XP_005681844.2 | 2 - 3 | 354 | 109 | |
| GenBank mRNA | JO602549.2 | - | 2 - 3 | 409 | 109 | |
More Information
Set Membership: |
|

CXCL11
Genomic Map