This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04733524_m1
See other ASIC1 GE Assays ›
Gene Symbol:
ASIC1
Gene Name:
acid sensing ion channel subunit 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 29463848 - 29488647 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04733524_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASIC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018047810.1 XP_017903299.1 7 - 8 1140 66
XM_018047811.1 XP_017903300.1 5 - 6 1581 66
GenBank mRNA JO632015.2 - 7 - 8 1183 66
JT690370.1 - 5 - 6 626 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0