Assay Details
| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041698.1 | XP_017897187.1 | 3 - 4 | 692 | 53 | |
| GenBank mRNA | JO428127.1 | - | 3 - 4 | 597 | 53 | |
More Information
Set Membership: |
|

ADGRA1
Genomic Map