Assay Details
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688319.3 | XP_005688376.1 | 4 - 5 | 502 | 54 | |
XM_005688320.3 | XP_005688377.1 | 4 - 5 | 502 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
