This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04737531_m1
See other GLYATL2 GE Assays ›
Gene Symbol:
GLYATL2
Gene Name:
glycine-N-acyltransferase like 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 1263731 - 1275441 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04737531_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLYATL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005690297.3 XP_005690354.1 3 - 4 464 91
XM_013969969.2 XP_013825423.1 3 - 4 636 91
GenBank mRNA KA344534.1 - 3 - 4 1057 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0