Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065645.1 | XP_017921134.1 | 2 - 3 | 304 | 91 | |
| GenBank mRNA | JO593574.2 | - | 5 - 6 | 582 | 91 | |
More Information
Set Membership: |
|

NEMP2
Genomic Map