This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04744060_mH
See other NUDT4 GE Assays ›
Gene Symbol:
NUDT4
Gene Name:
nudix hydrolase 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 22964341 - 22980669 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04744060_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005680075.3 XP_005680132.2 2 - 3 546 67
XM_018047622.1 XP_017903111.1 2 - 3 227 67
GenBank mRNA JO536695.2 - 3 - 4 456 67
JO632063.2 - 3 - 4 514 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0