Assay Details
| Gene Symbol: LOC102173207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058017.1 | XP_017913506.1 | 5 - 6 | 793 | 68 | |
| XM_018058018.1 | XP_017913507.1 | 5 - 6 | 790 | 68 | ||
More Information
Set Membership: |
|

LOC102173207
Genomic Map