Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694373.3 | XP_005694430.3 | 10 - 11 | 1910 | 93 | |
| XM_013972354.2 | XP_013827808.2 | 10 - 11 | 1205 | 93 | ||
| XM_018065400.1 | XP_017920889.1 | 10 - 11 | 1205 | 93 | ||
| GenBank mRNA | JO399391.2 | - | 2 - 3 | 270 | 93 | |
More Information
Set Membership: |
|

HEXD
Genomic Map