This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04751979_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.3: 42524644 - 42592530 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04751979_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005678287.3 XP_005678344.1 13 - 14 1486 74
XM_005678288.2 XP_005678345.1 14 - 15 1463 74
XM_005678289.2 XP_005678346.1 14 - 15 1566 74
XM_013962755.2 XP_013818209.1 12 - 13 1341 74
XM_013962756.2 XP_013818210.1 13 - 14 1347 74
XM_018045293.1 XP_017900782.1 15 - 16 1541 74
XM_018045307.1 XP_017900796.1 11 - 12 1117 74
XM_018045314.1 XP_017900803.1 14 - 15 1501 74
XM_018045319.1 XP_017900808.1 15 - 16 1584 74
XM_018045327.1 XP_017900816.1 12 - 13 1402 74
XM_018045337.1 XP_017900826.1 11 - 12 1258 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0