Assay Details
| Gene Symbol: SNAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687812.3 | XP_005687869.1 | 7 - 8 | 797 | 61 | |
| XM_005687813.3 | XP_005687870.1 | 7 - 8 | 797 | 61 | ||
| GenBank mRNA | JO634995.2 | - | 7 - 8 | 785 | 61 | |
More Information
Set Membership: |
|

SNAP25
Genomic Map