This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04757652_m1
See other NEIL1 GE Assays ›
Gene Symbol:
NEIL1
Gene Name:
nei like DNA glycosylase 1
Gene Aliases:
-
Chromosome Location:
Chr.21: 32908976 - 32915204 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04757652_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEIL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005695122.3 XP_005695179.3 1 - 2 618 64
XM_013973087.2 XP_013828541.2 1 - 2 619 64
XM_018066547.1 XP_017922036.1 3 - 4 634 64
XM_018066548.1 XP_017922037.1 2 - 3 613 64
XM_018066549.1 XP_017922038.1 2 - 3 910 64
GenBank mRNA JO593360.2 - 3 - 4 677 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline