This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04759869_m1
See other ARHGAP22 GE Assays ›
Gene Symbol:
ARHGAP22
Gene Name:
Rho GTPase activating protein 22
Gene Aliases:
-
Chromosome Location:
Chr.28: 2751852 - 2933544 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04759869_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018042511.1 XP_017898000.1 10 - 11 1413 65
XM_018042512.1 XP_017898001.1 8 - 9 1388 65
XM_018042513.1 XP_017898002.1 10 - 11 1364 65
XM_018042514.1 XP_017898003.1 8 - 9 1340 65
XM_018042515.1 XP_017898004.1 6 - 7 1069 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9bvz6:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline